Nalgene™ Narrow-Mouth HDPE IP2 Bottles, 4000mL - FAQs
View additional product information for Nalgene™ Narrow-Mouth HDPE IP2 Bottles - FAQs (2099-0001PK, 2099-0032, 2099-0010, 2099-0016PK, 2099-0032PK, 2099-0004PK, 2099-0010PK, 2099-0002PK, 2099-0008, 2099-0008PK, 2099-0016, 2099-0004, 2099-0001, 2099-0002)
1 product FAQs found
Are your Nalgene containers tested for air shipment of liquid samples?
Most of our products are manufactured and tested for leakproof performance in laboratory and industrial settings only. Our PassPort IP2 bottles (2099 and 2199 series) represent the only exception to this: they are rigorously leak tested under a 15 psi (-95 kPa) pressure differential to simulate unpressurized air cargo transport.