Dynabeads® HLA Class I (Not for sale in USA)

Catalog number:  21001D

 Related applications: Diagnostic Testing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

• Simple, effective, reliable Dynabeads antibody coated technology
• Isolation of peripheral blood T and B lymphocytes
• Isolates >99% pure and >95% viable cells within 15 minutes
• All isolation steps performed in a single tube
• No time consuming gradient centrifugation
• No costly equipment required

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols