FWS 제품은 특정 승인이 필요하므로 배송이 약 2-3주 더 소요됩니다.
Sf-900™ II SFM, no methionine, no cystine
Sf-900™ II SFM, no methionine, no cystine
Sf-900™ II SFM, no methionine, no cystine
Gibco™

Sf-900™ II SFM, no methionine, no cystine

Sf-900™ II SFM (without methionine and cystine) is a serum-free, protein-free insect cell culture medium optimized for the growth and자세히 알아보기
Have Questions?
카탈로그 번호수량
21012026500 mL
카탈로그 번호 21012026
제품 가격(KRW)
208,000
온라인 행사
Ends: 31-Dec-2026
244,000
할인액 36,000 (15%)
Each
수량:
500 mL
Customize this product
제품 가격(KRW)
208,000
온라인 행사
Ends: 31-Dec-2026
244,000
할인액 36,000 (15%)
Each
Sf-900™ II SFM (without methionine and cystine) is a serum-free, protein-free insect cell culture medium optimized for the growth and maintenance of Spodoptera frugiperda (Sf9 and Sf21) cells and for large-scale production of recombinant proteins expressed using the baculovirus expression vector system (BEVS). This medium is suitable for suspension and monolayer culture methods and supports growth of other lepidopteran cell lines. Gibco™ Sf-900™ II SFM features:

• Superior long-term, high-density growth
• Optimized for recombinant protein production
• Serum-free, protein-free, ready-to-use formulation
• Scalable in bioreactors

Superior long-term, high-density growth
Spodoptera frugiperda (Sf9) cells grown in Sf-900™ II SFM achieve maximum cell densities of 9 to 12 × 106 cells/mL, a significant improvement over competitors' formulations and Grace’s medium (see Product Manual). Increases in maximum cell densities of 20–100% are also observed with the Lymantria dispar (Gypsy moth) and Trichoplusia ni (Tn-368; Cabbage Looper) cell lines. Gibco™ Sf-900™ II SFM is capable of supporting the cultures to >20 passages.

Optimized for recombinant protein production
Traditionally, Grace's medium supplemented with 10% FBS has been used for recombinant protein expression. Sf-900™ II SFM is an improved serum-free, protein-free medium designed for growth of Sf9 and other lepidopteran cell lines and production of insect virus and rDNA proteins.

Serum-free, protein-free, ready-to-use formulation
Gibco™ Sf-900™ II SFM is a serum-free, protein-free medium that allows for much easier purification of your protein of interest. Sf-900™ II SFM is ready to use; it does not require addition of serum, glutamine, or surfactants. Cells adapted to other commercially available serum-free media can be subcultured directly into Sf-900™ II SFM, usually without any further adaptation. Cells usually require some adaptation from serum-containing formulations.

Scalable in bioreactors
The utility of Sf-900™ II SFM in larger-scale cell culture systems was demonstrated with a 5 L Celligen™ bioreactor. Successful infections with rAcNPV were carried out producing rβ-Gal and rEPO (see Product Manual).

Product use
Customers using Gibco™ Sf-900 II SFM in a manufacturing process, who have a submission with the FDA, may request a letter of authorization from us to reference our Type II Drug Master File (DMF).

cGMP manufacturing and quality system
For supply chain continuity, we manufacture Gibco™ Sf-900 II SFM at two separate facilities located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements, are certified to ISO 13485, and are registered with the FDA as medical device manufacturers.
For Research Use Only. Not for use in diagnostic procedures.
사양
세포주Sf21, Sf9
세포 유형Insect Cell
제품라인Gibco, Sf-900
제품 유형Insect Cell Serum Free Medium (SFM)
수량500 mL
배송 조건Room Temperature
종S. frugiperda, Spodoptera frugiperda
분류Protein-free, Serum-free
형태Liquid
Serum LevelSerum-free
Unit SizeEach
구성 및 보관
Storage conditions: 2°C to 8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

자주 묻는 질문(FAQ)

What antimicrobials can be used in insect culture and at what concentration?

Many antibiotics are suitable for use with insect cells. The following antibiotics are commonly used:
- Penicillin/Streptomycine: 50-100 U/mL; 50-100 µg/mL
- Amphotericin B (Fungizone antimycotic): 0.25 µg/mL
- Gentamicin: 0.5 mL of 10 mg/mL solution in 500 mL media (final concentration: 10 µg/mL)

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Is it necessary to heat-inactivate my serum before adding it to my medium?

Heat inactivation is not necessary. Our team has routinely used serum that has not been heat-inactivated, and we have not observed any effect on cell growth or morphology.

Many cells do not require heat-inactivated FBS. Some cells prefer heat-inactivated FBS. For instance, we use heat-inactivated FBS for our insect cell lines, i.e., Sf9 and Sf21 cells.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Do you offer serum-free insect media?

Yes, we offer a variety of serum-free insect media. Please go here (http://www.thermofisher.com/us/en/home/life-science/cell-culture/insect-cell-culture/insect-cell-culture-misc/serum-free-media.html?icid=cvc-insect-media-c2t2) to view the media we offer and the differences between them.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.