1,2-Dimethyl-5-nitroimidazole, 98%
1,2-Dimethyl-5-nitroimidazole, 98%
1,2-Dimethyl-5-nitroimidazole, 98%
Thermo Scientific Chemicals

1,2-Dimethyl-5-nitroimidazole, 98%

CAS: 551-92-8 | C5H7N3O2 | 141.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Catalog number 210330010
Price (USD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS551-92-8
IUPAC Name1,2-dimethyl-5-nitro-1H-imidazole
Molecular FormulaC5H7N3O2
InChI KeyIBXPYPUJPLLOIN-UHFFFAOYSA-N
SMILESCN1C(C)=NC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point137°C to 142°C
Appearance (Form)Crystalline powder
Appearance (Color)Cream to yellow
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only