Hexanes, technical, mixture of isomers
Hexanes, technical, mixture of isomers
Hexanes, technical, mixture of isomers
Hexanes, technical, mixture of isomers
Thermo Scientific Chemicals

Hexanes, technical, mixture of isomers

CAS: 92112-69-1 | C6H14
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 210590025
Price (USD)
-
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS92112-69-1
IUPAC Namehexane
Molecular FormulaC6H14
InChI KeyVLKZOEOYAKHREP-UHFFFAOYSA-N
SMILESCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Appearance (Form)Clear liquid
Residue after evaporation=<0.002 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only