2,2-Diethoxyacetophenone, 96%
2,2-Diethoxyacetophenone, 96%
2,2-Diethoxyacetophenone, 96%
Thermo Scientific Chemicals

2,2-Diethoxyacetophenone, 96%

CAS: 6175-45-7 | C12H16O3 | 208.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
Glass bottle
Catalog number 210620050
Price (USD)
25.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
25.00
Each
Chemical Identifiers
CAS6175-45-7
IUPAC Name2,2-diethoxy-1-phenylethan-1-one
Molecular FormulaC12H16O3
InChI KeyPIZHFBODNLEQBL-UHFFFAOYSA-N
SMILESCCOC(OCC)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to dark yellow
Appearance (Form)Liquid
Refractive index1.4980 to 1.5010 (20°C, 589 nm)
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only