Cobalt(II) acetylacetonate, 99%
Cobalt(II) acetylacetonate, 99%
Cobalt(II) acetylacetonate, 99%
Thermo Scientific Chemicals

Cobalt(II) acetylacetonate, 99%

CAS: 14024-48-7 | C10H14CoO4 | 257.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number 210640500
Price (USD)
98.20
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
98.20
Each
Chemical Identifiers
CAS14024-48-7
IUPAC Nameλ²-cobalt(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC10H14CoO4
InChI KeyUTYYEGLZLFAFDI-FDGPNNRMSA-L
SMILES[Co++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Purple
Appearance (Form)Crystalline powder
Assay98.5 to 101.5 % (ex Co)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only