3-Chloro-2,4-pentanedione, 98%
3-Chloro-2,4-pentanedione, 98%
3-Chloro-2,4-pentanedione, 98%
Thermo Scientific Chemicals

3-Chloro-2,4-pentanedione, 98%

CAS: 1694-29-7 | C5H7ClO2 | 134.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Catalog number 210675000
Price (USD)
-
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS1694-29-7
IUPAC Name3-chloropentane-2,4-dione
Molecular FormulaC5H7ClO2
InChI KeyVLRGXXKFHVJQOL-UHFFFAOYSA-N
SMILESCC(=O)C(Cl)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to brown
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4825 to 1.4845 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only