4-(Dimethylamino)phenethyl alcohol, 99%
4-(Dimethylamino)phenethyl alcohol, 99%
4-(Dimethylamino)phenethyl alcohol, 99%
Thermo Scientific Chemicals

4-(Dimethylamino)phenethyl alcohol, 99%

CAS: 50438-75-0 | C10H15NO | 165.236 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 210930010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS50438-75-0
IUPAC Name2-[4-(dimethylamino)phenyl]ethan-1-ol
Molecular FormulaC10H15NO
InChI KeyCDTPAAZQBPSVGS-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(CCO)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to light brown
GC>=98.5 %
Appearance (Form)Crystalline powder
Melting point53°C to 58°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only