Orion™ 2109XP Fluoride Analyzer
El producto real puede variar
Thermo Scientific™

Orion™ 2109XP Fluoride Analyzer

Maximize uptime and keep your plant in perfect operation by offering unmatched versatility and performance with our fluoride analyzer.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipo
2109XPAnalizador de fluoruro
210960XPKit de consumibles de fluoruro
210960XPFKit de consumibles F sin reactivos
2109XPCALKit de calibración de fluoruro
2109XPEKKit de electrodos de fluoruro
2109XPENAnalizador con carcasa
2109XPEPPlaca frontal de componentes electrónicos
2109XPSKKit de servicio
Número de catálogo 2109XP
Precio (MXN)
-
Tipo:
Analizador de fluoruro
Maximize up-time and keep your plant in pristine operation with the versatility and performance of the Thermo Scientific™ Orion™ 2109XP Fluoride Analyzer. It offers accuracy, reliability and ease of use to best meet the demands of your fluoride analysis.

Features:

  • Uses a recognized EPA method for fluoride analysis using an ion selective electrode (ISE)
  • Eliminates problems with turbidity and suspended solids in samples
  • Provides true accuracy in measuring total fluoride, with a detection limit of 10ppb
  • Offers continuous fluoride monitoring in a wide variety of samples
  • Protects against over-fluoridation of samples with sensitive, selective, reliable and verifiable measurements
  • Provides early detection of fluoride
  • Limits unnecessary calibration cycles due to drift with superior Orion fluoride electrode technology
  • Minimizes operator time and maintenance without the use of complicated moving parts or pumps that often require frequent attention and expensive spare part expenditures

Advanced User Interface:

  • Detailed calibration, measurement and diagnostic logs for early diagnostic and action level notification
  • Configurable to your facility's desired level of performance
  • Password protected
  • Easy to install, allowing your plant to be up and running in minutes

Options:

  • Retrofitable to the Orion 1809AO panel mounting footprint
  • Available in several configurations including the analyzer instrument only or with a protective enclosure

Periodo de garantía limitado: Este producto está sujeto a todos los requisitos de la garantía y a las exclusiones establecidas por nuestros términos y condiciones de venta. El plazo de garantía comienza a partir de la fecha del envío del producto y finaliza tras noventa (90) días. Exención de responsabilidad de la restricción de uso para investigación interna: cualquier restricción impuesta al comprador para utilizar el producto o productos únicamente para fines de investigación interna no se aplica al producto o productos que aparecen en las páginas web asociadas a esta declaración.

Especificaciones
Exactitud±10% or 10 ppb, whichever is greater
Exactitud (temperatura)±0.5°C
Exactitud (mV)± (0.5 mV + 0.1%)
Salida de alarma3
Amperaje5A
Salidas analógicas2, Galvanically Isolated
CalibraciónDouble Known Addition
Material del envaseValox 364
Certificaciones/conformidadCE, CSA, UL, FCC Class A limits
Channel 1Fluoride
Channel 2Optional ph/ORP or Conductivity
Canales1
Detalles de comunicaciónModbus with Additional Module
Bucles de corriente2
Descripción2109XP Fluoride Analyzer
Dimensiones (Largo x Ancho x Alto)11 x 17 x 26 in. (27 x 45 x 65cm)
Tipo de pantallaCustom backlit LCD with temperature, concentration, mV, error codes and menu driven prompts
Clasificación de carcasaWaterproof; IP66; NEMA 4X
Caudal40mL/min.
Frecuencia0 to 60 Hz
Altura (imperial)26 in.
Altura (métrico)65 cm
IncluyeFluoride Analyzer Only
Presión de entrada8 to 100 psig
Entradas85-132; 170-264 VAC
Tipo de tecladoMembrane
Longitud (imperial)11 in.
Longitud (métrico)27 cm
Humedad relativa máx.5 to 95%, non-condensing
Intervalos de medición10 ppb to 200 ppm
Opciones de montajeWall mount
Salidas0 to 20 mA; 4 to 20 mA
Detalles del relé3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Tiempo de respuesta95% within 2 minutes of injecting a standard solution
Entrada de muestras1/4 in. NPTF tube fitting
Flujos de muestras1
Tensión85/132 VAC, 170/264 VAC
Anchura (imperial)17 in.
Anchura (métrico)45 cm
Temperatura de funcionamiento (métrico)5°C to 35°C
TipoAnalizador de fluoruro
Unit SizeEach