Orion™ 2109XP Fluoride Analyzer
Actual product may vary
Thermo Scientific™
Orion™ 2109XP Fluoride Analyzer
Maximize uptime and keep your plant in perfect operation by offering unmatched versatility and performance with our fluoride analyzer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
2109XPCALFluoride Calibration Kit
210960XPFluoride Consumables Kit
210960XPFReagentless F- Consumables Kit
2109XPFluoride Analyzer
2109XPEKFluoride Electrode Kit
2109XPENAnalyzer with Enclosure
2109XPEPElectronics Faceplate
2109XPSKService Kit
Catalog number 2109XPCAL
Price (USD)
1,287.00
Online exclusive
1,430.00
Save 143.00
Each
Type:
Fluoride Calibration Kit
Price (USD)
1,287.00
Online exclusive
1,430.00
Save 143.00
Each
Maximize up-time and keep your plant in pristine operation with the versatility and performance of the Thermo Scientific™ Orion™ 2109XP Fluoride Analyzer. It offers accuracy, reliability and ease of use to best meet the demands of your fluoride analysis.
Features:
  • Uses a recognized EPA method for fluoride analysis using an ion selective electrode (ISE)
  • Eliminates problems with turbidity and suspended solids in samples
  • Provides true accuracy in measuring total fluoride, with a detection limit of 10ppb
  • Offers continuous fluoride monitoring in a wide variety of samples
  • Protects against over-fluoridation of samples with sensitive, selective, reliable and verifiable measurements
  • Provides early detection of fluoride
  • Limits unnecessary calibration cycles due to drift with superior Orion fluoride electrode technology
  • Minimizes operator time and maintenance without the use of complicated moving parts or pumps that often require frequent attention and expensive spare part expenditures
Advanced User Interface:
  • Detailed calibration, measurement and diagnostic logs for early diagnostic and action level notification
  • Configurable to your facility's desired level of performance
  • Password protected
  • Easy to install, allowing your plant to be up and running in minutes
Options:
  • Retrofitable to the Orion 1809AO panel mounting footprint
  • Available in several configurations including the analyzer instrument only or with a protective enclosure
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Alarm Output3
Analog Outputs2109XPCAL
Case MaterialValox 364
Certifications/Compliance2109XPCAL
DescriptionFluoride Calibration Kit with Carrying Case
Enclosure RatingWaterproof; IP66; NEMA 4X
Flow Rate40 mL/min.
Includes2109XPCAL
Inlet Pressure8 to 100 psig
Inputs85-132; 170-264 VAC
Outputs0-20; 4-20 mA
Relay Details3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Response Time95% within 2 minutes of injecting a standard solution
Sample Inlet1/4 NPTF tube fitting in.
Temperature (Metric) Operating5°C to 35°C
TypeFluoride Calibration Kit
Unit SizeEach