Orion™ 2109XP Fluoride Analyzer
Orion™ 2109XP Fluoride Analyzer
El producto real puede variar
Thermo Scientific™
Orion™ 2109XP Fluoride Analyzer
Maximize uptime and keep your plant in perfect operation by offering unmatched versatility and performance with our fluoride analyzer.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipo
2109XPSKKit de servicio
210960XPKit de consumibles de fluoruro
210960XPFKit de consumibles F sin reactivos
2109XPAnalizador de fluoruro
2109XPCALKit de calibración de fluoruro
2109XPEKKit de electrodos de fluoruro
2109XPENAnalizador con carcasa
2109XPEPPlaca frontal de componentes electrónicos
Número de catálogo 2109XPSK
Precio (MXN)
-
Tipo:
Kit de servicio
Maximize up-time and keep your plant in pristine operation with the versatility and performance of the Thermo Scientific™ Orion™ 2109XP Fluoride Analyzer. It offers accuracy, reliability and ease of use to best meet the demands of your fluoride analysis.
Features:
  • Uses a recognized EPA method for fluoride analysis using an ion selective electrode (ISE)
  • Eliminates problems with turbidity and suspended solids in samples
  • Provides true accuracy in measuring total fluoride, with a detection limit of 10ppb
  • Offers continuous fluoride monitoring in a wide variety of samples
  • Protects against over-fluoridation of samples with sensitive, selective, reliable and verifiable measurements
  • Provides early detection of fluoride
  • Limits unnecessary calibration cycles due to drift with superior Orion fluoride electrode technology
  • Minimizes operator time and maintenance without the use of complicated moving parts or pumps that often require frequent attention and expensive spare part expenditures
Advanced User Interface:
  • Detailed calibration, measurement and diagnostic logs for early diagnostic and action level notification
  • Configurable to your facility's desired level of performance
  • Password protected
  • Easy to install, allowing your plant to be up and running in minutes
Options:
  • Retrofitable to the Orion 1809AO panel mounting footprint
  • Available in several configurations including the analyzer instrument only or with a protective enclosure
Periodo de garantía limitado: Este producto está sujeto a todos los requisitos de la garantía y a las exclusiones establecidas por nuestros términos y condiciones de venta. El plazo de garantía comienza a partir de la fecha del envío del producto y finaliza tras noventa (90) días. Exención de responsabilidad de la restricción de uso para investigación interna: cualquier restricción impuesta al comprador para utilizar el producto o productos únicamente para fines de investigación interna no se aplica al producto o productos que aparecen en las páginas web asociadas a esta declaración.
Especificaciones
Salida de alarma3
Salidas analógicas2109XPSK
Material del envaseValox 364
Certificaciones/conformidad2109XPSK
Descripción2109XP Service Kit
Clasificación de carcasaWaterproof; IP66; NEMA 4X
Caudal40 mL/min.
Incluyeelectronics faceplate (2109XPEP), air pump assembly (2100PA), regulator assembly (2100RG), 2 electrode cables with gland assemblies for sensing and reference electrodes (2100EC), power supply assembly (2100PS) and fuse kit assembly (2100FK115 and 2100FK230)
Presión de entrada8 to 100 psig
Entradas85-132; 170-264 VAC
Salidas0-20; 4-20 mA
Detalles del relé3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Tiempo de respuesta95% within 2 minutes of injecting a standard solution
Entrada de muestras1/4 NPTF tube fitting in.
Temperatura de funcionamiento (métrico)5°C to 35°C
TipoKit de servicio
Unit SizeEach