Nalgene™ Square Wide-Mouth PPCO Bottles with Closure
Actual product may vary
Nalgene™ Square Wide-Mouth PPCO Bottles with Closure
Nalgene™ Square Wide-Mouth PPCO Bottles with Closure
Nalgene™ Square Wide-Mouth PPCO Bottles with Closure
Nalgene™ Square Wide-Mouth PPCO Bottles with Closure
Nalgene™ Square Wide-Mouth PPCO Bottles with Closure
Nalgene™ Square Wide-Mouth PPCO Bottles with Closure

Nalgene™ Square Wide-Mouth PPCO Bottles with Closure

Simplify cleaning, sample filling and retrieval with these space-efficient, wide-mouth PPCO bottles, which are contact-clear, autoclavable and resistant to most acids, alcohols and bases.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeUnit Size
2110-0006PK
also known as 2110-0006
175 mL38 mmPack of 12
2110-000260 mL28 mmCase of 72
2110-0002PK
also known as 2110-0002
60 mL28 mmPack of 12
2110-0006175 mL38 mmCase of 72
2110-0008250 mL43 mmCase of 72
2110-0008PK
also known as 2110-0008
250 mL43 mmPack of 12
2110-0016500 mL53 mmCase of 48
2110-0016PK
also known as 2110-0016
500 mL53 mmPack of 12
2110-00321 L63 mmCase of 24
2110-0032PK
also known as 2110-0032
1 L63 mmPack of 6
Catalog number 2110-0006PK
also known as 2110-0006
Price (USD)
56.09
Online exclusive
65.98
Save 9.89
Pack of 12
Capacity (Metric):
175 mL
Closure Size:
38 mm
Unit Size:
Pack of 12
Price (USD)
56.09
Online exclusive
65.98
Save 9.89
Pack of 12

Save valuable lab space with the square design of Thermo Scientific™ Nalgene™ Wide-Mouth Square Polypropylene Copolymer Bottles. Wide-mouth design allows easy cleaning, sample filling and retrieval. Made from only high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables.

Features:
  • Resistant to acids, alcohols and bases
  • Contact-clear bottle provides visible liquid level
  • Square design provides a large area for labeling
  • Linerless polypropylene closures provide leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
  • Autoclavable
  • Leakproof† when used with Nalgene closures
  • Manufactured from non-toxic materials

Note:

Completely disengage threads or remove cap before autoclaving.

.

Recycling Information:
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  These bottles and their packaging are recyclable in many communities. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene PP and PPCO bottles if services are available in your community.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
AutoclavableAutoclavable
Capacity6 oz., 175mL
Capacity (English)6 oz.
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size38 mm
Closure TypeScrew Cap
DescriptionWide-Mouth Square Bottle
Diameter (Metric)52.1 mm
Height (English)4.17 in.
Height (Metric)105.9 mm
Product TypeSquare Wide Mouth Bottle
ShapeSquare
Through the Closure DispensingNo
Capacity (Metric)175 mL
Closure IncludedYes
ColorTranslucent
GraduatedNo
MaterialPPCO
MouthWide
SterilityNon-sterile
Unit SizePack of 12

Frequently asked questions (FAQs)

Are the Nalgene PPCO media bottles/tubes able to be irradiated?

PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.