D-Gluconic acid, calcium salt, 99%
D-Gluconic acid, calcium salt, 99%
D-Gluconic acid, calcium salt, 99%
Thermo Scientific Chemicals

D-Gluconic acid, calcium salt, 99%

CAS: 299-28-5 | C12H22CaO14 | 430.372 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 211060010
Price (USD)
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS299-28-5
IUPAC Namecalcium bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
Molecular FormulaC12H22CaO14
InChI KeyNEEHYRZPVYRGPP-IYEMJOQQSA-L
SMILES[Ca++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration after Ion exchange98.5 to 102.0 % (as C12H22CaO14.H2O)
Loss on drying=<2.0 % (1 g, 105°C, 16 hrs)
Sulfate (SO4)=<500 ppm
Appearance (Color)White to off-white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only