Consumables and Services Kits for Orion™ 2110XP Ammonia Analyzer
Actual product may vary
Thermo Scientific™

Consumables and Services Kits for Orion™ 2110XP Ammonia Analyzer

Keep needed supplies and service items in stock to avoid lost productivity with Thermo Scientific™ Consumables and Services Kits for Orion™ 2110XP Ammonia Analyzer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
2110XPEKAmmonia Electrode Kit
210056Reference Electrode Half-Cell
211050XPAmmonia Consumables Kit
211050XPFReagentless Consumables Kit
2110XPCALAmmonia Calibration Kit
2110XPEPElectronics Faceplate
2110XPSKService Kit
Catalog number 2110XPEK
Price (USD)
-
Type:
Ammonia Electrode Kit
Keep needed supplies and service items in stock to avoid lost productivity with Thermo Scientific™ Consumables and Services Kits for the Thermo Scientific™ Orion™ 2110XP Ammonia Analyzer.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Accuracy (Temperature)±0.5 °C
Accuracy (mV)±(0.5 mV + 0.1%)
DescriptionAmmonia Electrode Kit - includes ammonia electrode, reference electrode and 2 oz. reference electrode filling solution
Display Type2110XPEK
Flow Rate40 mL/min.
Includesammonia electrode (100047), reference electrode (210056) and 2 oz. reference electrode filling solution (181073)
Inlet Pressure8 to 100 psig
Inputs85-132; 170-264 VAC
OutputsAlarm: 3; Relay: 3; Analog: 2; Selections: 0-20mA; 4-20 mA
Range (mV)±1999.9 m
Relay DetailsMax Load: 250VAC; 30VDC; 5A
Resolution (Temperature)0.1 °C
Resolution (mV)0.1 mV
Response TimeInitial response within 15 seconds, reach 90% of reading within 1 minute of injecting a standard solution
Sample Inlet0.25 NPTF Tube Fitting in.
Sample Temperature5 to 45 °C
Temperature (Metric) Operating-10°C to +120°C
TypeAmmonia Electrode Kit
Unit SizeEach