Silver hexafluorophosphate, 98%, -6 mesh
Silver hexafluorophosphate, 98%, -6 mesh
Silver hexafluorophosphate, 98%, -6 mesh
Thermo Scientific Chemicals

Silver hexafluorophosphate, 98%, -6 mesh

CAS: 26042-63-7 | AgF6P | 252.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
25 g
Packaging:
Plastic bottle
Catalog number 211120100
Price (USD)
-
Quantity:
10 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS26042-63-7
IUPAC Namesilver(1+) hexafluoro-λ⁵-phosphanuide
Molecular FormulaAgF6P
InChI KeySCQBROMTFBBDHF-UHFFFAOYSA-N
SMILES[Ag+].F[P-](F)(F)(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Silver (Ag)>=41.6 %
Appearance (Color)White to grey-beige
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only