Orion™ 2111XP Sodium Analyzer
Actual product may vary
Thermo Scientific™

Orion™ 2111XP Sodium Analyzer

Meet the demanding needs of high-purity water measurements and high-acid cation exchange applications with the Thermo Scientific™ Orion™ 2111XP Sodium Analyzer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
2111XPENNa Analyzer, Enclosure
2111XPNa Analyzer
2111XPENGNa Analyzer, Encl, Grab Sampler
2111XPGNa Analyzer, Grab Sampler
Catalog number 2111XPEN
Price (INR)
-
Type:
Na Analyzer, Enclosure
Meet the demanding needs of high-purity water measurements and high-acid cation exchange applications with the Thermo Scientific™ Orion™ 2111XP Sodium Analyzer. This system offers unmatched reliability in analyzing critical sample streams throughout the power/steam generation and industrial water industry. It is an all-in-one system from one of the most trusted names in sodium monitoring.
  • Provides early warning detection of sodium
  • Features simple, step-by-step scrolling instruction
  • Offers measurements at a glance with visible backlit display
  • Limits calibration cycles with Orion sodium electrode technology
  • Minimizes of operator time and maintenance without use of complicated moving parts or pumps
  • Detects limit of 0.1ppb
  • Offers flexible application packages suited for continuous sodium monitoring in a wide variety of samples

Ordering Information:

  • Calibration kit, Cat. No. 2111XPCAL
  • Calibration standards, Cat. No. 181140 and 181141v
  • Sodium electrode, Cat. No. 210045
  • Reference electrode, Cat. No. 210056
  • Reference electrode filling solution, Part No. 181073

Ammonia Application:

  • Application package, Cat. No. 2111XA
  • Annual consumables Kit, Cat. No. 211160XP
  • Reagent, Cat. No. 181130
  • Diffusion tubing, Cat. No. 150060

DIPA Application:

  • Application package, Cat. No. 2111XD
  • Annual consumables kit, Cat. No. 211192XP
  • Reagent, Cat. No. 211190
  • Diffusion tubing, Cat. No. 211194

Cation/High Acid Application:

  • Application package, Cat. No. 2111XC
  • Annual consumables kit, Cat. No. 211150XP
  • Reagent, Cat. No. 181130
  • Diffusion tubing, Cat. No. 181160
Specifications
Accuracy±5% or 0.1 ppb, whichever is greater
Accuracy (Temperature)±0.5°C
Accuracy (mV)± (0.5 mV + 0.1%)
Alarm Output3
Amperage5A
Analog Outputs2, Galvanically Isolated
CalibrationDouble Known Addition
Case MaterialValox 364
Certifications/ComplianceCE, CSA, UL, FCC Class A limits
Channel 1Sodium
Channel 2Optional ph/ORP or Conductivity
Channels1
Communication DetailsModbus with Additional Module
Current Loops2
Description2111XP Sodium Analyzer With Enclosure
Detection Limits0.1 ppb
Dimensions (L x W x H)11 x 17 x 26 in. (27 x 45 x 65 cm)
Display TypeCustom backlit LCD with temperature, concentration, mV, error codes and menu driven prompts
Enclosure RatingWaterproof; IP66; NEMA 4X
Flow Rate40 mL/min.
Frequency0 to 60 Hz
Height (English)26 in.
Height (Metric)65 cm
Includes2111XP sodium analyzer with Enclosure, sodium electrode, reference electrode, ATC probe, sodium calibration standards kits, CD user guide and options kit
Inlet Pressure8 to 100 psig
Keypad TypeMembrane
Length (English)11 in.
Length (Metric)27 cm
Max. Relative Humidity5 to 95%, non-condensing
Measurement Ranges0 to 10 ppm
Mounting OptionsWall mount
Outputs0 to 20 mA; 4 to 20 mA
Relay Details3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Response Time95% within 2 minutes of injecting a standard solution
Sample Inlet1/4 in. NPTF tube fitting
Sample Streams1
Sample Temperature5°C to 45°C
Voltage85/132 VAC, 170/264 VAC
Width (English)17 in.
Width (Metric)45 cm
Temperature (Metric) Operating5°C to 35°C
TypeNa Analyzer, Enclosure
Unit SizeEach