Nalgene™ PPCO Mason Jars with Closure, 500mL - FAQs
Nalgene™ PPCO Mason Jars with Closure, 500mL - FAQs
View additional product information for Nalgene™ PPCO Mason Jars with Closure - FAQs (2115-1000, 2115-2000, 2115-0500, 2115-1000PK, 2115-2000PK, 2115-0500PK, 2115-3000PK, 2115-3000)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.