Bacto™ Neopeptone
Gibco™

Bacto™ Neopeptone

Bacto Neopeptone is an animal origin (AO) enzymatic digest of bovine (milk) and porcine protein. This nutritional supplement provides vitamins,Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
21168010 kg
211681500 g
Catalog number 211680
Price (USD)
10,760.00
Each
Quantity:
10 kg
Price (USD)
10,760.00
Each
Bacto Neopeptone is an animal origin (AO) enzymatic digest of bovine (milk) and porcine protein. This nutritional supplement provides vitamins, nucleotides, minerals, and peptides in a variety of sizes to any culture formulation. Customers have used Bacto Neopeptone as a component of culture media in the cultivation of human pathogens, notably Bordetella pertussis and group A streptococci, as well as Borrelia burgdorferi and Clostridium species, to produce vaccines for animal health applications.

Features:
• Enzymatic digest of bovine (milk) and porcine protein
• Animal origin nutritional supplement
• Contains a wide variety of peptide sizes in combination with vitamins, nucleotides, and minerals

Applications of Bacto Neopeptone include:
• Recommended for use in media for detection of fungi
• Reported to provide nutrients for support of spirochetes and protozoa
• Microbial fermentation
• Production of vaccines for animal health applications

Specifications
For Use With (Application)Production of vaccines for animal health applications
Product LineBacto
Quantity10 kg
SourceAnimal origin, bovine and porcine
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.