Bacto Tryptone is an animal origin (AO) pancreatic digest of casein. It is included in standard methods manual applications andRead more
Have Questions?
Change view
Catalog Number
Quantity
211699
2 kg
211701
10 kg
211705
500 g
3 Options
Catalog number 211699
Price (USD)
764.00
Each
Quantity:
2 kg
Price (USD)
764.00
Each
Bacto Tryptone is an animal origin (AO) pancreatic digest of casein. It is included in standard methods manual applications and is listed in the Reagent section of the United States Pharmacopeia as meeting the specifications for a pancreatic digest of casein, a component in many of the media listed. The nutritional profile of this supplement is unique in that it provides a range of nutrients. Customers have used this supplement in fermentation applications and shown that it successfully supports the growth of many bacterial species during the production of animal health vaccines, including Escherichia coli, and Fusobacterium and Agrobacterium species when used together with Bacto Yeast Extract.
Features: • Pancreatic digest of casein • Developed while investigating a peptone particularly suitable for the elaboration of indole by bacteria • Animal origin nutritional supplement
Applications of Bacto Tryptone include: • Works well in fermentation applications • Use with common organisms such as Escherichia coli, as well as uncommon organisms such as the diatom Nitzschia laevis
Specifications
Certifications/ComplianceLUO, MUO
For Use With (Application)Dehydrated culture media
FormatPail
Product LineBacto
Quantity2 kg
Shelf Life1825 Days
SourceAnimal Origin
Product TypeBacterial Media
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Frequently asked questions (FAQs)
What alternative product do you recommend for Bacto Tryptone (Cat. No. 211699, 211701, 211705)?
We have several casein digests that could serve as alternatives to Bactone Tryptone (Cat. No. 211699, 211791, 211705), including Bacto Casitone (Cat. No. 225930, 225910), Difco Tryptone Peptone (Cat. No. 21921, 211922, 211923), BiTek Tryptone (Cat. No. 251420). However, the digestion and nutritive profile of each peptone are different, therefore there may be differences in bacterial growth or supplementation ability. We recommend empirically determining the performance of alternative casein digestion in your application.