Thioflavin T
Thioflavin T
Thioflavin T
Thermo Scientific Chemicals

Thioflavin T

CAS: 2390-54-7 | C17H19ClN2S | 318.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 211760250
Price (USD)
114.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
114.00
Each
Chemical Identifiers
CAS2390-54-7
IUPAC Name2-[4-(dimethylamino)phenyl]-3,6-dimethyl-1,3-benzothiazol-3-ium chloride
Molecular FormulaC17H19ClN2S
InChI KeyJADVWWSKYZXRGX-UHFFFAOYSA-M
SMILES[Cl-].CN(C)C1=CC=C(C=C1)C1=[N+](C)C2=CC=C(C)C=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to golden to orange-brown
UV Lambda max409 to 415 nm (10 mg/l in water)
Solubility(1 mg/ml in methanol) Clear to slightly hazy yellow to yellow-green
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only