Orion™ 2117LL Low-Level Chloride Analyzer
El producto real puede variar
Orion™ 2117LL Low-Level Chloride Analyzer
Orion™ 2117LL Low-Level Chloride Analyzer
Thermo Scientific™
Orion™ 2117LL Low-Level Chloride Analyzer
Get continuous online monitoring and control of steam cycle and boiler chemistry with the Thermo Scientific™ Orion™ 2117LL Low-Level Chloride Analyzer.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipoTensión
2117LLEN230Analizador de cloruro, carcasa230 V
2117LLAnalizador de cloruro110 V
2117LLENAnalizador de cloruro, carcasa110 V
2117LL230Analizador de cloruro230 V
Número de catálogo 2117LLEN230
Precio (MXN)
-
Tipo:
Analizador de cloruro, carcasa
Tensión:
230 V
Get continuous online monitoring and control of steam cycle and boiler chemistry with the Thermo Scientific™ Orion™ 2117LL Low-Level Chloride Analyzer, for use with power generation applications requiring chloride measurement at trace levels.
Without accurate and real-time monitoring of chloride, the risk of corrosion fatigue, stress corrosion cracking and pitting can create significant costs in maintenance and downtime. Early detection of chloride ingress from feedwater helps your plant maintain the optimal balance of water purification with ease and peace of mind.
Features:
  • Detects chloride in real-time, providing accurate and reliable results
  • Provides high performance results with confidence
  • Includes unique electrode technology for stability and a virtually maintenance-free fluidic design
Benefits:
  • Calibration record is maintained for up to 60 days between reagent changes.
  • System is one of the only real-time monitors on the market capable of reading down to 5ppb.
  • Long-life, solid-state sensor technology offers stability and high performance.
  • No moving parts means simple operation and minimal maintenance.
  • Measurements are repeatable and verifiable.
Ordering information:
The chloride analyzer is available in several configurations including the analyzer instrument only or with a protective enclosure.
Especificaciones
DescripciónLow Level Chloride Analyzer with Enclosure, 230V
Tensión230 V
TipoAnalizador de cloruro, carcasa
Unit SizeEach