Orion™ 2117XP Chloride Analyzer
Actual product may vary
Thermo Scientific™

Orion™ 2117XP Chloride Analyzer

Detect chloride in real time with accurate and reliable results using the Thermo Scientific™ Orion™ 2117XP Chloride Analyzer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
2117XPENProtective Enclosure Package
2117XPInstrument Only Package
Catalog number 2117XPEN
Price (ZAR)
-
Includes:
Protective Enclosure Package
Detect chloride in real time with accurate and reliable results using the Thermo Scientific™ Orion™ 2117XP Chloride Analyzer. Early detection of chloride ingress from feedwater helps your plant maintain the optimal balance of water purification and peace of mind.
Features:
  • Detection limit of 0.1ppm offers continuous chloride monitoring in wide variety of samples.
  • Protection against early breakthrough with sensitive, selective, reliable and verfiable measurements provides early detection of chloride in RO and boiler systems.
  • Fast and highly accurate calibration has you back online in minutes.
  • System holds calibration up to 60 days between reagent changes.
  • Long life solid state sensor technology offers stability and high performance.
  • Simple operation and no moving parts means only minimal maintenance is required.
  • Get repeatable and verifiable measurements at a glance using the large, user-friendly display.

2117XP Chloride Analyzer Ordering Information:

  • Annual Consumables Kit, Cat. No. 211750XP
  • Reagent, Cat. No. 182011
  • Diffusion Tubing, Cat. No. 181060
  • Calibration Kit, Cat. No. 2117XPCAL
  • Calibration Standards, Cat. No. 211740
  • Chloride Electrode, Cat. No. 100025
  • Reference Electrode, Cat. No. 100057
  • Reference Electrode Filling Solution, Cat. No. 100057F4
Specifications
Accuracy±10% or 0.1 ppm, whichever is greater
Accuracy (Temperature)±0.5°C
Accuracy (mV)± (0.5 mV + 0.1%)
Alarm Output3
Amperage5A
Analog Outputs2, Galvanically Isolated
CalibrationDouble Known Addition
Case MaterialValox 364
Certifications/ComplianceCE, CSA, UL, FCC Class A limits
Channel 1Chloride
Channel 2Optional ph/ORP or Conductivity
Channels1
Communication DetailsModbus with Additional Module
Current Loops2
Description2117XP Chloride Analyzer With Enclosure
Detection Limits0.1ppm
Dimensions (L x W x H)11 x 17 x 26 in. (27 x 45 x 65cm)
Display TypeCustom backlit LCD with temperature, concentration, mV, error codes and menu driven prompts
Enclosure RatingIP66, NEMA 4X
Flow Rate40mL/min.
Frequency0 to 60 Hz
Height (English)26 in.
Height (Metric)65 cm
IncludesProtective Enclosure Package
Inlet Pressure8 to 100psig
Keypad TypeMembrane
Length (English)11 in.
Length (Metric)27 cm
Max. Relative Humidity5 to 95%, non-condensing
Measurement Ranges0 to 10ppm
Mounting OptionsWall mount
Outputs0 to 20 mA; 4 to 20 mA
Relay Details3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Response Time95% within 2 minutes of injecting a standard solution
Sample Flow Rate40 mL/min nominal set by pressure regulator
Sample Inlet1/4 in. NPTF tube fitting
Sample Streams1
Voltage85/132 VAC, 170/264 VAC
Width (English)17 in.
Width (Metric)45 cm
Temperature (Metric) Operating5°C to 35°C
TypeChloride Analyzer
Unit SizeEach