Gelysate™ Peptone
Gibco™

Gelysate™ Peptone

Gelysate Peptone is a pancreatic digest of gelatin, derived from porcine animal origin (AO) sources. Gelatin hydrolysate is high in proline residues. Gelysate Peptone is deficient in carbohydrates and is characterized by low cystine, methionine, and tryptophan content.
Have Questions?
Catalog NumberQuantity
211870454 g
Catalog number 211870
Price (USD)
156.00
Each
Quantity:
454 g
Price (USD)
156.00
Each

Gelysate Peptone is a pancreatic digest of gelatin, derived from porcine animal origin (AO) sources. Gelatin hydrolysate is high in proline residues. Gelysate Peptone is deficient in carbohydrates and is characterized by low cystine, methionine, and tryptophan content.

Specifications
AdditivePeptone
Certifications/ComplianceLUO
For Use With (Application)Formulation of culture media for the cultivation of heterotrophic microorganisms
Packaging TypeBottle
Product LineGelysate
Quantity454 g
Shelf Life1460 Days
FormSolid
Unit SizeEach
Contents & Storage
Store at 2°C to 25°C in cool, dry conditions in well-sealed containers.