Polypeptone Peptone is a mixture of peptones made up of equal parts of pancreatic digest of casein and peptic digestRead more
Have Questions?
Catalog Number
Quantity
211910
454 g
Catalog number 211910
Price (USD)
229.00
Each
Quantity:
454 g
Price (USD)
229.00
Each
Polypeptone Peptone is a mixture of peptones made up of equal parts of pancreatic digest of casein and peptic digest of animal tissue. Polypeptone Peptone includes the high content of amino acids and small polypeptides characteristic of pancreatic digest of casein and the larger polypeptides characteristic of peptic digest of animal tissue.
Features: • Animal origin mixture of peptones • Includes the high content of amino acids
Applications of Polypeptone Peptone include: • Found to meet nutritional requirements of various bacteria, fungi, and mammalian cells for which a single source of casein or meat peptones has been unsatisfactory
Specifications
Certifications/ComplianceLUO
For Use With (Application)Various bacteria, fungi, and mammalian cells
FormatBottle
Product LinePolypeptone
Quantity454 g
Shelf Life1460 Days
SourceAnimal Origin
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.