Orion™ 2120XP Calcium Hardness Analyzer
El producto real puede variar
Thermo Scientific™

Orion™ 2120XP Calcium Hardness Analyzer

Limit the costly effects of improper water purification using the Thermo Scientific™ Orion™ 2120XP Calcium Hardness Analyzer.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoClasificación de carcasa
2120XPCarcasa no incluida
2120XPENIP66, NEMA 4X
Número de catálogo 2120XP
Precio (MXN)
-
Clasificación de carcasa:
Carcasa no incluida
Limit the costly effects of improper water purification using the Thermo Scientific™ Orion™ 2120XP Calcium Hardness Analyzer. This analyzer detects calcium as a major component of hardness in real time, providing accurate and reliable results.
Features:
  • Holds calibration up to 60 days between reagent changes
  • Fast and accurate calibration to get you back online in minutes
  • Long life sensor technology for stability and high performance
  • Simple operation and minimal maintenance required—no moving parts
  • Repeatable and verifiable measurements at a glance using the large, user friendly display

2120XP Calcium Hardness Analyzer Ordering Information:

  • Annual Consumables Kit, Cat. No. 212050XP
  • Reagent, Cat. No. 182011
  • Diffusion Tubing, Cat. No. 181060
  • Calibration Kit, Cat. No. 2120XPCAL
  • Calibration Standards, Cat. No. 182040
  • Calcium Electrode, Cat. No. 100049
  • Reference Electrode, Cat. No. 100056
  • Reference Electrode Filling Solution, Cat. No. 181073

Recommended for:

  • Water softening
  • Zeolite softener effluent monitoring
  • Blending applications
Especificaciones
Exactitud±10% or 10 ppb, whichever is greater
Exactitud (temperatura)±0.5°C
Exactitud (mV)± (0.5 mV + 0.1%)
Salida de alarma3
Amperaje5A
Salidas analógicas2, Galvanically Isolated
CalibraciónDouble Known Addition
Material del envaseValox 364
Certificaciones/conformidadCE, CSA, UL, FCC Class A limits
Channel 1Calcium
Channel 2Optional ph/ORP or Conductivity
Canales1
Detalles de comunicaciónModbus with Additional Module
Bucles de corriente2
Descripción2120XP Calcium Hardness Analyzer
Dimensiones (Largo x Ancho x Alto)11 x 17 x 26 in. (27 x 45 x 65 cm)
Tipo de pantallaCustom backlit LCD with temperature, concentration, mV, error codes and menu driven prompts
Clasificación de carcasaCarcasa no incluida
Caudal40 mL/min.
Frecuencia0 to 60 Hz
Altura (imperial)26 in.
Altura (métrico)65 cm
Incluye2120XP calcium analyzer with calcium electrode, reference electrode, ATC probe, calcium calibration standards kit, CD user guide and options kit
Presión de entrada8 to 100 psig
Tipo de tecladoMembrane
Longitud (imperial)11 in.
Longitud (métrico)27 cm
Humedad relativa máx.5 to 95%, non-condensing
Intervalos de medición0 to 10ppm
Opciones de montajeWall mount
Salidas0 to 20 mA; 4 to 20 mA
Detalles del relé3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Tiempo de respuesta95% within 2 minutes of injecting a standard solution
Caudal de la muestra40 mL/min nominal set by pressure regulator
Entrada de muestras1/4 in. NPTF tube fitting
Flujos de muestras1
Temperatura de funcionamiento (imperial)41°F to 95°F
Tensión85/132 VAC, 170/264 VAC
Anchura (imperial)17 in.
Anchura (métrico)45 cm
Temperatura de funcionamiento (métrico)5°C to 35°C
TipoCalcium Hardness Analyzer
Unit SizeEach