Bacto™ Proteose Peptone No. 2
Gibco™

Bacto™ Proteose Peptone No. 2

Bacto Proteose Peptone No. 2 is member of the Bacto Proteose Peptones, which are enzymatic digests of protein. Studies ofRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
21211010 kg
212120500 g
Catalog number 212110
Price (USD)
11,030.00
Each
Quantity:
10 kg
Price (USD)
11,030.00
Each
Bacto Proteose Peptone No. 2 is member of the Bacto Proteose Peptones, which are enzymatic digests of protein. Studies of peptic digests of animal tissue prepared under varying digestion parameters demonstrated that no single peptone is the most suitable nitrogen source for every microbiological application. These findings led to the development of Bacto Proteose Peptone, Bacto Proteose Peptone No. 2, and Bacto Proteose Peptone No. 3. Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone.

• Bacto Proteose Peptone is used in preparing microbiological culture media and in producing bacterial toxins
• Bacto Proteose Peptone No. 2 is used in preparing microbiological culture media
• Bacto Proteose Peptone No. 3 is used in preparing microbiological and mammalian cell culture media
• Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone. It offers the same beneficial nutrients as Bacto Proteose Peptone for growth promotion and toxin production in a wide range of fastidious microorganisms.

We also offer alternatives to Bacto Proteose Peptones for scale-up to production applications, including BiTek Proteose Peptone.
Specifications
Culture TypeMammalian, Insect, Microbial
For Use With (Application)Formulation of culture media for the cultivation of heterotrophic microorganisms
Product LineBacto
Quantity10 kg
SourceAnimal Origin
FormPowder
Product TypeBacterial Media
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.