Indigo, synthetic
Indigo, synthetic
Indigo, synthetic
Thermo Scientific Chemicals

Indigo, synthetic

CAS: 482-89-3 | C16H10N2O2 | 262.268 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 212131000
Price (USD)
100.65
Online exclusive
112.00
Save 11.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
100.65
Online exclusive
112.00
Save 11.35
Each
Chemical Identifiers
CAS482-89-3
IUPAC Name(Z)-1H,1'H,3H,3'H-[2,2'-biindolylidene]-3,3'-dione
Molecular FormulaC16H10N2O2
InChI KeyCOHYTHOBJLSHDF-YPKPFQOOSA-N
SMILESO=C1\C(NC2=CC=CC=C12)=C1\NC2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark blue to violet
UV spectrumComplies
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only