Clean-Blot™ IP Detection Reagent (HRP) - FAQs
View additional product information for Clean-Blot™ IP Detection Reagent (HRP) - FAQs (21230)
1 product FAQs found
How does the Clean-Blot IP Detection Kit work?
This assay uses a HRP conjugate or AP conjugate that binds to native IgG from various host species, allowing clear, specific Western blot detection from IP experiments without interference from denatured IgG. It is used simply as a substitute for secondary antibodies.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.