Product Image
Gibco™

Beef Extract Powder

Beef Extract Powder is a meat extract dried into a powder. This nutritional supplement provides a mixture of peptides, individual amino acids, nucleotides, organic acids, minerals, and vitamins.
Have Questions?
Catalog NumberQuantity
212303500 g
Catalog number 212303
Price (CLP)
100.026
Each
Quantity:
500 g
Price (CLP)
100.026
Each

Beef Extract Powder is a meat extract dried into a powder. This nutritional supplement provides a mixture of peptides, individual amino acids, nucleotides, organic acids, minerals, and vitamins. Additionally, due to differences in the process, Beef Extract Powder has a different molecular weight profile than other beef extract products in our portfolio. Customers have used Beef Extract Powder in a variety of microbial fermentation cell lines including Clostridium sp., Escherichia coli, Bacillus subtillis, Pasteurella multocida, and Streptococcus species to produce numerous animal health vaccines.

Features:

  • An animal origin (AO) nutritional supplement
  • A mixture of peptides, individual amino acids, nucleotides, organic acids, minerals, and vitamins

Applications of Beef Extract Powder include:

  • Used in a variety of microbial fermentation cell lines
  • Used to produce numerous animal health vaccines
Specifications
AdditiveBeef Extract
Certifications/ComplianceAOAC, COMPF, SMWW, ISO/DIS, 11014
For Use With (Application)Dehydrated culture media
Packaging TypeBottle
Quantity500 g
Shelf Life1460 Days
FormPowder
Unit SizeEach
Contents & Storage
Store at 2°C to 25°C in cool, dry conditions in well-sealed containers.