Product Image
Gibco™

Difco™ Beef Extract

Difco Beef Extract is an animal origin peptone derived from an infusion of beef. This nutritional supplement provides a mixture of peptides, individual amino acids, nucleotides, organic acids, minerals, and vitamins.
Have Questions?
Catalog NumberQuantity
212610500 g
Catalog number 212610
Price (USD)
286.00
Each
Quantity:
500 g
Price (USD)
286.00
Each

Difco Beef Extract is an animal origin peptone derived from an infusion of beef. This nutritional supplement provides a mixture of peptides, individual amino acids, nucleotides, organic acids, minerals, and vitamins. Provided as a paste, customers have used Difco Beef Extract in a variety of microbial fermentation cell lines, including Escherichia coli, Bacillus subtillis, Pasteurella multocida, and Streptococcus species to produce numerous animal health vaccines.

Features:

  • An animal origin nutritional supplement
  • A mixture of peptides, individual amino acids, nucleotides, organic acids, minerals, and vitamins

Applications of Difco Beef Extract include:

  • Use in a variety of microbial fermentation cell lines
  • Used to produce numerous animal health vaccines
Specifications
Certifications/ComplianceLUO
For Use With (Application)Cultivation of heterotrophic microorganisms
FormatBottle
Product LineDifco
Quantity500 g
Shelf Life1460 Days
SourceAnimal origin-free
FormPaste
Product TypeBacterial Media
Unit SizeEach
Contents & Storage
Store at 2°C to 25°C in cool, dry conditions in well-sealed containers.