Isopropylmagnesium chloride, 2.0M solution in THF, AcroSeal™
Isopropylmagnesium chloride, 2.0M solution in THF, AcroSeal™
Isopropylmagnesium chloride, 2.0M solution in THF, AcroSeal™
Thermo Scientific Chemicals

Isopropylmagnesium chloride, 2.0M solution in THF, AcroSeal™

CAS: 1068-55-9 | C3H7ClMg | 102.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number 212851000
Price (USD)
67.70
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)
67.70
Each
Chemical Identifiers
CAS1068-55-9
IUPAC Namechloro(propan-2-yl)magnesium
Molecular FormulaC3H7ClMg
InChI KeyGPIBKUJXKCEZOH-UHFFFAOYSA-M
SMILESCC(C)[Mg]Cl
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay develop some turbidity or precipitate
Appearance (Color)Colorless to yellow to grey to dark brown
Appearance (Form)Solution
Titration Grignard1.80 to 2.20 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only