Cobalt(II) sulfate heptahydrate, 99+%, extra pure
Cobalt(II) sulfate heptahydrate, 99+%, extra pure
Cobalt(II) sulfate heptahydrate, 99+%, extra pure
Cobalt(II) sulfate heptahydrate, 99+%, extra pure
Thermo Scientific Chemicals

Cobalt(II) sulfate heptahydrate, 99+%, extra pure

CAS: 10026-24-1 | CoH14O11S | 281.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 213105000
Price (USD)
289.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
289.00
Each
Chemical Identifiers
CAS10026-24-1
IUPAC Nameλ²-cobalt(2+) heptahydrate sulfate
Molecular FormulaCoH14O11S
InChI KeyMEYVLGVRTYSQHI-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red-brown
Titration Complexometric>=99.0 %
Appearance (Form)Adhering crystalline powder or crystals
Nickel (Ni)=<500 ppm
Lead (Pb)=<50 ppm
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only