EZ-Link™ NHS-PEG12-Biotin, 500 mg - FAQs
View additional product information for EZ-Link™ NHS-PEG12-Biotin, No-Weigh™ Format - FAQs (21313, 21312, A35389)
2 product FAQs found
Can you provide the shelf-life for EZ-Link NHS-PEG12-Biotin?
EZ-Link NHS-PEG12-Biotin is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (-20 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What is the advantage of using a PEGylated NHS biotinylation reagent?
The hydrophilic polyethylene glycol (PEG) spacer arm imparts water solubility that is transferred to the biotinylated molecule. For example, antibodies labeled with NHS-PEG4- Biotin exhibit less aggregation when stored in solution as compared to antibodies labeled with reagents having only hydrocarbon spacers. Specific labeling of cell surface proteins is another useful application for these water-soluble and membrane-impermeable reagents.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.