Sodium ethoxide, pure, 21% in ethanol
Sodium ethoxide, pure, 21% in ethanol
Sodium ethoxide, pure, 21% in ethanol
Thermo Scientific Chemicals

Sodium ethoxide, pure, 21% in ethanol

CAS: 141-52-6 | C2H5NaO | 68.051 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 213222500
Price (USD)
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS141-52-6
IUPAC Namesodium ethanolate
Molecular FormulaC2H5NaO
InChI KeyQDRKDTQENPPHOJ-UHFFFAOYSA-N
SMILES[Na+].CC[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Yellow to brown
Titration with HCl20.0 to 22.0 %
AppearanceMay darken on storage
NaOH + Na2CO3 (as NaOH)=<0.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use