EZ-Link™ Sulfo-NHS-LC-Biotin - FAQs
View additional product information for EZ-Link™ Sulfo-NHS-LC-Biotin - FAQs (21335B)
1 product FAQs found
What is the difference between EZ-Link Sulfo-NHS-LC-Biotin and Sulfo-NHS-LC-LC-Biotin?
EZ-Link Sulfo-NHS-LC-Biotin has a spacer arm length of 22.4 Angstroms while EZ-Link Sulfo-NHS-LC-LC-Biotin has a spacer arm length of 30.5 Angstroms.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.