Nitromethane, 99+%, for analysis
Nitromethane, 99+%, for analysis
Nitromethane, 99+%, for analysis
Thermo Scientific Chemicals

Nitromethane, 99+%, for analysis

CAS: 75-52-5 | CH3NO2 | 61.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 213370025
Price (USD)
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS75-52-5
IUPAC Namenitromethane
Molecular FormulaCH3NO2
InChI KeyLYGJENNIWJXYER-UHFFFAOYSA-N
SMILESC[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Lead (Pb)=<0.1 ppm
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only