What are the differences between Pierce Streptavidin Magnetic Beads and MagnaBind Streptavidin Beads?
Pierce Streptavidin Magnetic Beads are homogeneous, non-aggregating, pre-blocked, iron oxide, superparamagnetic microparticles encapsulated with a proprietary polymer surface (no exposed iron, bead size of approximately 1 (0.5-1.5) µm, concentration 10 mg/mL). Pierce Streptavidin Magnetic Beads can be used in manual or automated applications (using KingFisher 96 and KingFisher Flex instruments, for example).
MagnaBind Streptavidin Beads are also superparamagnetic particles, but are heterogeneous aggregates made of silanized iron oxide (bead size approximately 1-4 µm, concentration 5 mg/mL). MagnaBind Streptavidin Beads can only be used in manual applications.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
MagnaBind Streptavidin Beads are also superparamagnetic particles, but are heterogeneous aggregates made of silanized iron oxide (bead size approximately 1-4 µm, concentration 5 mg/mL). MagnaBind Streptavidin Beads can only be used in manual applications.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.