MagnaBind™ Goat Anti-Rabbit IgG
Thermo Scientific™

MagnaBind™ Goat Anti-Rabbit IgG

Enable benchtop magnetic affinity purification with these ligand-coated (streptavidin, Protein A and G, GAM IgG, GAR IgG) magnetic particles and magnet separators.
Have Questions?
카탈로그 번호수량
2135650 mL
카탈로그 번호 21356
제품 가격(KRW)
603,000
온라인 행사
Ends: 30-Sep-2026
689,000
할인액 86,000 (12%)
Each
수량:
50 mL
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
603,000
온라인 행사
Ends: 30-Sep-2026
689,000
할인액 86,000 (12%)
Each

Thermo Scientific™ MagnaBind™ Beads provide a convenient method for magnetic separation of antibodies, antigens, lectins, enzymes, nucleic acids and cells using affinity binding. To remove the MagnaBind Beads from the suspension, an external magnetic field is used.

MagnaBind Goat Anti-Rabbit IgG Beads can be used to conveniently separate cells of interest from a cell mixture. The monoclonal or polyclonal antibody to the cell surface antigen is pre-incubated with the appropriate magnetic bead that is then incubated with the cell suspension.

Features of MagnaBind Goat Anti-Rabbit IgG Beads:

  • Composition: Silanized iron oxide
  • Magnetization: 25-35EMU/g
  • Type of Magnetization: Superparamagnetic (no magnetic memory)
  • Surface Area: >100m2/g
  • Bead Size: 1-4μm diameter
  • Settling Rate: 4% in 30 minutes
  • Effective Density: 2.5g/mL
  • Number of Beads: 1 x 108 beads/mg
  • pH Stability: Aqueous solution, above pH 4.0
  • Concentration: ∼1 mg/mL

Applications:

  • Cell sorting using either positive or negative selection
  • Protein purification or immunoassays using either direct or indirect methods
For Research Use Only. Not for use in diagnostic procedures.
사양
농도Slurry: ∼1 mg/mL
설명MagnaBind Goat Anti-Rabbit IgG Beads
리간드 유형Antibody
제품라인MagnaBind
수량50 mL
부피(미터법)50 mL
색상Brown
직경(미터법)1 to 4 μm
용도(애플리케이션)Affinity Purification
제품 유형Antibody Coated Bead
Unit SizeEach
구성 및 보관
Upon receipt store products at 4°C. Do not freeze products. Products are shipped at ambient temperature.