Tetrachloro-p-benzoquinone, 99%
Tetrachloro-p-benzoquinone, 99%
Tetrachloro-p-benzoquinone, 99%
Acros Organics

Tetrachloro-p-benzoquinone, 99%

CAS: 118-75-2 | C6Cl4O2 | 245.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
250 g
Packaging:
Plastic bottle
Catalog number 213565000
also known as 21356
HSN 98020000
Price (INR)
39,934.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
39,934.00
Each
Chemical Identifiers
CAS118-75-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-green
Appearance (Form)Powder or crystals
Infrared spectrumConforms
HPLC>=98.5 %
Loss on drying=<0.5 % (1 g, 105°C, 3 hrs)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only