Molybdic acid, for analysis
Molybdic acid, for analysis
Molybdic acid, for analysis
Thermo Scientific Chemicals

Molybdic acid, for analysis

CAS: 7782-91-4 | H2MoO4 | 161.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number 213621000
Price (USD)
71.65
Special offer
Online exclusive
Ends: 28-Dec-2026
83.70
Save 12.05
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
71.65
Special offer
Online exclusive
Ends: 28-Dec-2026
83.70
Save 12.05
Each
Chemical Identifiers
CAS7782-91-4
IUPAC Namedioxomolybdenumdiol
Molecular FormulaH2MoO4
InChI KeyVLAPMBHFAWRUQP-UHFFFAOYSA-L
SMILESO[Mo](O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Additional infocontains ammonium molybdate
Appearance (Form)Powder
Titration with NaOH>=85 % (as MoO3)
Heavy metals (as Pb)=<30 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only