4-Pentynoic acid, 95%
4-Pentynoic acid, 95%
4-Pentynoic acid, 95%
Thermo Scientific Chemicals

4-Pentynoic acid, 95%

CAS: 6089-09-4 | C5H6O2 | 98.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 213730010
Price (USD)
57.65
Special offer
Online exclusive
Ends: 28-Dec-2026
67.90
Save 10.25
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
57.65
Special offer
Online exclusive
Ends: 28-Dec-2026
67.90
Save 10.25
Each
Chemical Identifiers
CAS6089-09-4
IUPAC Namepent-4-ynoate
Molecular FormulaC5H5O2
InChI KeyMLBYLEUJXUBIJJ-UHFFFAOYSA-M
SMILES[O-]C(=O)CCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or flakes and/or chunks
Melting point54°C to 60°C
Infrared spectrumConforms
Appearance (Color)White to yellow to beige to light brown
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only