ART™ Barrier Pipette Tips, TipCycle Recycle Kit, Hinged Racks
ART™ Barrier Pipette Tips, TipCycle Recycle Kit, Hinged Racks
ART™ Barrier Pipette Tips, TipCycle Recycle Kit, Hinged Racks
ART™ Barrier Pipette Tips, TipCycle Recycle Kit, Hinged Racks
ART™ Barrier Pipette Tips, TipCycle Recycle Kit, Hinged Racks
ART™ Barrier Pipette Tips, TipCycle Recycle Kit, Hinged Racks
Thermo Scientific™

ART™ Barrier Pipette Tips, TipCycle Recycle Kit, Hinged Racks

The pipette tip and rack mail-back program provides a simple and effective solution for recycling PP#5 single use plastics. The TipCycle Kit included with your case of pipette tips to collect used pipette tips and empty racks. Complete waiver, attach pre-paid shipping label and return for recycling.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Tip StyleModel
2139RC-HR10 μLStandardART 10 μL, Barrier Sterile Hinged Rack, TipCycle Recycle Kit
2065ERC-HR100 μLStandardART 100 μL, Barrier Sterile Hinged Rack, TipCycle Recycle Kit
2069RC-05-HR200 μLLow RetentionART 200 μL, Low Retention, Barrier Sterile Hinged Rack, TipCycle Recycle Kit
2139RC-05-HR10 μLLow RetentionART 10 μL, Low Retention, Barrier Sterile Hinged Rack, TipCycle Recycle Kit
2149PRC-05-HR20 μLLow RetentionART 20 μL, Low Retention, Barrier Sterile Hinged Rack, TipCycle Recycle Kit
2179RC-HR1000 μLStandardART 1000 μL, Barrier Sterile Hinged Rack, TipCycle Recycle Kit
2069RC-HR200 μLStandardART 200 μL, Barrier Sterile Hinged Rack, TipCycle Recycle Kit
2149PRC-HR20 μLStandardART 20 μL, Barrier Sterile Hinged Rack, TipCycle Recycle Kit
2065ERC-05-HR100 μLLow RetentionART 100 μL, Low Retention, Barrier Sterile Hinged Rack, TipCycle Recycle Kit
Catalog number 2139RC-HR
Price (USD)
-
Volume (Metric):
10 μL
Tip Style:
Standard
Model:
ART 10 μL, Barrier Sterile Hinged Rack, TipCycle Recycle Kit

Unlike standard length barrier tips, Thermo Scientific™ ART™ Barrier Extended Length Pipette Tips in Hinged Racks self-seal when exposed to potential contaminants. The pipette tips from the reload inserts fit securely into the environmentally friendly, reusable hinged racks. The locking clasp prevents contamination by keeping remaining tips sterile and offers an easy, one-handed operation. Available in MicroPoint™, low retention and offer a universal fit design.

Features:

  • Recycle nonhazardous pipette tips, racks, and associated packaging with Tipcycle
  • Complete TipCycle Pipette Tip Recycling Kit Provides: 1 case of pipette tips, instructions for return, a box liner and liner tie help prevent liquid spills, decontamination waiver must be completed and inserted into recycle package, FedEx return shipping label adhered to top of recycle package, then simply ship the recycle package
  • Stackable hinged racks save space in your lab
  • Sterile tips certified free: DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin

Recommended for:

  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
Racks offer quick and easy access to pipette tips, saving valuable time
  • Front latch closes and locks, ensuring safe and secure storage throughout
  • Flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
  • Secure snap design securely holds insert in place, preventing it from being lifted out while pipetting
  • Lid rim prevents racks from sliding around when stacked and carted around a lab
Specifications
Certifications/ComplianceISO 9001:2015
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
DisposableSingle-use
For Use With (Application)Liquid Handling, Molecular Biology, PCR
GraduatedNo
Includes1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Length (English)1.23 in.
Length (Metric)3.13 cm
ModelART 10 μL, Barrier Sterile Hinged Rack, TipCycle Recycle Kit
Product TypeTipCycle, Pipette Tip and Rack Recycling Kit
Quantity4800/Cs.
Volume (Metric) Dispensing10 μL
AutoclavableNot Autoclavable
ColorNatural
FilteredFiltered
Fit for PurposeSterile and PCR Ready
For Use With (Equipment)Universal Fit
FormatHinged Rack
MaterialPolypropylene
Product LineART
SterilitySterile
Tip StyleStandard
Volume (Metric)10 μL
Unit SizeCase of 4800