Acetonitrile-d3, for NMR, contains 1 v/v% TMS, 99 atom % D
Acetonitrile-d3, for NMR, contains 1 v/v% TMS, 99 atom % D
Acetonitrile-d3, for NMR, contains 1 v/v% TMS, 99 atom % D
Thermo Scientific Chemicals

Acetonitrile-d3, for NMR, contains 1 v/v% TMS, 99 atom % D

CAS: 2206-26-0 | C2H3N | 44.07 g/mol
数量:
10 mL
50 mL
パッケージ:
Glass Bottle
製品番号(カタログ番号) 214530100
価格(JPY)
-
数量:
10 mL
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2206-26-0
IUPAC Name(²H₃)acetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-FIBGUPNXSA-N
SMILES[2H]C([2H])([2H])C#N
さらに表示
Appearance (Color)Clear colorless
D-Enrichment>=98.5 %
Appearance (Form)Liquid
Water=<0.1 % (Coulometric)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only