Sodium hexanitrocobaltate(III)
Sodium hexanitrocobaltate(III)
Sodium hexanitrocobaltate(III)
Thermo Scientific Chemicals

Sodium hexanitrocobaltate(III)

CAS: 13600-98-1 | CoN6Na3O12 | 403.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 214861000
Price (USD)
158.65
Special offer
Online exclusive
Ends: 28-Dec-2026
186.00
Save 27.35
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
158.65
Special offer
Online exclusive
Ends: 28-Dec-2026
186.00
Save 27.35
Each
Chemical Identifiers
CAS13600-98-1
IUPAC Nametrisodium hexanitrocobalttris(ylium)
Molecular FormulaCoN6Na3O12
InChI KeyBZFWEBJDBATXME-UHFFFAOYSA-N
SMILES[Na+].[Na+].[Na+].[O-][N+](=O)[Co+3]([N+]([O-])=O)([N+]([O-])=O)([N+]([O-])=O)([N+]([O-])=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to orange-brown
Appearance (Form)Powder
Suitability(for K determination) Passes test
Insoluble matter=<0.02 % (in dilute acetic acid)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only