HEPES sodium salt, 99%
HEPES sodium salt, 99%
HEPES sodium salt, 99%
Thermo Scientific Chemicals

HEPES sodium salt, 99%

CAS: 75277-39-3 | C8H17N2NaO4S | 260.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
1 kg
Catalog number 215000250
Price (USD)
58.60
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
58.60
Each
Chemical Identifiers
CAS75277-39-3
IUPAC Namesodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
Molecular FormulaC8H17N2NaO4S
InChI KeyRDZTWEVXRGYCFV-UHFFFAOYSA-M
SMILES[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with HClO4>=98.5 % (on anhydrous substance)
pH9.5 to 10.5 (1 % in water)
UVat 280 nm A: =<0.02
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only