Spermidine trihydrochloride, 99+%
Spermidine trihydrochloride, 99+%
Spermidine trihydrochloride, 99+%
Thermo Scientific Chemicals

Spermidine trihydrochloride, 99+%

CAS: 334-50-9 | C7H22Cl3N3 | 254.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 215100050
Price (USD)
120.65
Online exclusive
134.00
Save 13.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
120.65
Online exclusive
134.00
Save 13.35
Each
Chemical Identifiers
CAS334-50-9
IUPAC Nametrihydrogen (4-aminobutyl)(3-aminopropyl)amine trichloride
Molecular FormulaC7H22Cl3N3
InChI KeyLCNBIHVSOPXFMR-UHFFFAOYSA-N
SMILES[H+].[H+].[H+].[Cl-].[Cl-].[Cl-].NCCCCNCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration Argentometric>=99.0 %
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only