Rhodamine B base, 97%
Rhodamine B base, 97%
Rhodamine B base, 97%
Thermo Scientific Chemicals

Rhodamine B base, 97%

Rhodamine B base, CAS # 509-34-2, is a fluorescent dye used in staining applications. | CAS: 509-34-2 | C28H30N2O3
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 215230250
Price (USD)
52.65
Online exclusive
58.10
Save 5.45
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
52.65
Online exclusive
58.10
Save 5.45
Each
Chemical Identifiers
CAS509-34-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink to red to violet
UV Lambda max542 to 546 nm (in methanol)
Appearance (Form)Powder and/or chunks
Dye content>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Rhodamine B is an organic water-soluble fluorescent dye
  • Rhodamine B has been found as a toxic pollutant in aqueous systems that may be removed by photocatalysis methods

Applications

  • Rhodamine B has been used to prepare fluorescent probes for iron detection
  • It can be used for fluorescent labeling of nanoparticles and subsequent detection of nanoparticle uptake in cells
  • Is used as a dye in ELISA, fluorescence microscopy, and flow cytometry
RUO – Research Use Only

General References:

  1. Tahir, H.; Saad, M. Chapter 3 - Using dyes to evaluate the photocatalytic activity. Ed. Mehrorang Ghaedi,
  2. In: Interface Science and Technology, Elsevier, 32 (2021) pp 125-224.
  3. Benyettou, F; Chebbi, I; Motte, L; and Seksek, O. Magnetoliposome for alendronate delivery. J. Mater. Chem., 2011,21, 4813-4820
  4. Qin, Z.; Su, W.; Liu, P.; Ma, J.; Zhang, Y.; Jiao, T. Facile Preparation of a Rhodamine B Derivative-Based Fluorescent Probe for Visual Detection of Iron Ions. ACS Omega. 2021, 6(38), 25040-25048.