Benzyl cinnamate, 99%
Benzyl cinnamate, 99%
Benzyl cinnamate, 99%
Thermo Scientific Chemicals

Benzyl cinnamate, 99%

CAS: 103-41-3 | C16H14O2 | 238.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 215291000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS103-41-3
IUPAC Namebenzyl (2E)-3-phenylprop-2-enoate
Molecular FormulaC16H14O2
InChI KeyNGHOLYJTSCBCGC-VAWYXSNFSA-N
SMILESO=C(OCC1=CC=CC=C1)\C=C\C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline mass or liquid after melting
Melting point33°C to 37°C
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only