1-Eicosanol, 98%
1-Eicosanol, 98%
1-Eicosanol, 98%
Thermo Scientific Chemicals

1-Eicosanol, 98%

CAS: 629-96-9 | C20H42O | 298.56 g/mol
Have Questions?
Catalog NumberQuantityPackaging
2154600505 gGlass bottle
Catalog number 215460050
Price (USD)
31.65
Special offer
Online exclusive
Ends: 28-Dec-2026
37.40
Save 5.75
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
31.65
Special offer
Online exclusive
Ends: 28-Dec-2026
37.40
Save 5.75
Each
Chemical Identifiers
CAS629-96-9
IUPAC Nameicosan-1-ol
Molecular FormulaC20H42O
InChI KeyBTFJIXJJCSYFAL-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)White
Melting point62°C to 66°C
Appearance (Form)Powder or pellets and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only